3,5-Dibromo-4,6-dimethylpyridin-2-amine 95+%

Supplier: Ambeed
Warning

A506797-100MG A506797-1G A506797-250MG
77017-260EA 58 USD
77017-260 77017-262 77017-264
3,5-Dibromo-4,6-dimethylpyridin-2-amine 95+%
3,5-Dibromo-4,6-dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₈Br₂N₂
MDL Number: MFCD00975063
CAS Number: 5407-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR