Ethyl 4-(1-Imidazolyl)benzoate 98%

Supplier: Ambeed
Warning

A506782-1G A506782-250MG
77004-738EA 25 USD
77004-738 77004-740
Ethyl 4-(1-Imidazolyl)benzoate 98%
Ethyl 4-(1-Imidazolyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD03425368
CAS Number: 86718-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR