3-Bromo-2-chloro-6-methylpyridine 98%

Supplier: Ambeed
Warning

A506566-100G A506566-10G A506566-1G A506566-25G A506566-5G
77004-728EA 227 USD
77004-728 77004-730 77004-732 77004-734 77004-736
3-Bromo-2-chloro-6-methylpyridine 98%
3-Bromo-2-chloro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MDL Number: MFCD03095218
CAS Number: 185017-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR