Ethyl 3-methylpyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

A506456-100MG A506456-1G
77017-256EA 26.53 USD
77017-256 77017-258
Ethyl 3-methylpyrazine-2-carboxylate 95%
Ethyl 3-methylpyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD09839066
CAS Number: 25513-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR