H-p-Nitro-D-Phe-OMe·HCl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Phe(4-NO₂)-OMe·HCl, H-D-Phe(4-NO2)-OMe·HCl, H-D-Phe(4-NO2)-OMe.HCl, H-p-Nitro-D-Phe-OMe·HCl
A506117-1G
A506117-250MG
A506117-5G
A506117-25G
77017-240EA
13.8
USD
77017-240
77017-242
77017-244
77684-327
H-p-Nitro-D-Phe-OMe·HCl 98%
H-p-Nitro-D-Phe-OMe·HCl
Precaución: For research use only.
Formula:
C₁₀H₁₂N₂O₄·HCl MW: 260.674 g/mol |
MDL Number:
MFCD00797553 CAS Number: 67877-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...