4-Methoxy-N-phenylbenzamide 95+%

Supplier: Ambeed
Warning

A506107-100MG A506107-1G A506107-250MG
76991-310EA 28.65 USD
76991-310 76991-312 76991-314
4-Methoxy-N-phenylbenzamide 95+%
4-Methoxy-N-phenylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.263 g/mol
MDL Number: MFCD00025810
CAS Number: 7465-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR