1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine

A505614-1G A505614-25G A505614-5G
77034-534EA 6.37 USD
77034-534 77034-536 77034-538
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine 98%
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₄FN₃O₂
MW: 239.25 g/mol
MDL Number: MFCD02633484
CAS Number: 221198-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR