3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride 97%

Supplier: Ambeed
Warning

A505587-100G A505587-10G A505587-1G A505587-250MG A505587-25G A505587-5G
76991-294EA 594.11 USD
76991-294 76991-296 76991-298 76991-300 76991-302 76991-304
3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride 97%
3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNOS
MW: 205.71 g/mol
MDL Number: MFCD23703103
CAS Number: 645411-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR