5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A505119-1G
A505119-250MG
A505119-5G
77004-718EA
37.14
USD
77004-718
77004-720
77004-722
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile 95%
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₁₃N₃ |
MDL Number:
MFCD11846194 CAS Number: 269726-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...