5-(Methylsulfonyl)nicotinic acid 96%

Supplier: Ambeed
Warning

A504804-100MG A504804-1G A504804-250MG
77034-528EA 32 USD
77034-528 77034-530 77034-532
5-(Methylsulfonyl)nicotinic acid 96%
5-(Methylsulfonyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₄S
MDL Number: MFCD06254799
CAS Number: 893723-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR