4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Danger

A504754-100MG A504754-1G
77004-714EA 26.53 USD
77004-714 77004-716
4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine 97%
4,5-Dichloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD12401653
CAS Number: 1142192-58-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR