6-Acetyl-2-Benzoxazolinone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Acetyl-2-benzoxazolinone, 6-Acetyl-2(3H)-benzoxazolone, 6-Acetyl-2-hydroxybenzoxazole, 6-Acetyl-2-benzoxazolone, 6-Acetylbenzo[d]oxazol-2(3H)-one

A504307-1G A504307-5G
76991-280EA 28 USD
76991-280 77886-297
6-Acetyl-2-Benzoxazolinone 98%
6-Acetyl-2-Benzoxazolinone

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD01664312
CAS Number: 54903-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR