3-Chloro-5-(trifluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A504085-1G A504085-250MG A504085-25G
76991-274EA 22 USD
76991-274 76991-276 76991-278
3-Chloro-5-(trifluoromethoxy)benzoic acid 97%
3-Chloro-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₃
MDL Number: MFCD06660306
CAS Number: 433926-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR