Methyl 5-bromo-2-(dimethylamino)benzoate 95%

Supplier: Ambeed
Warning

A503947-250MG A503947-5G
76991-270EA 68 USD
76991-270 76991-272
Methyl 5-bromo-2-(dimethylamino)benzoate 95%
Methyl 5-bromo-2-(dimethylamino)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
MDL Number: MFCD11110844
CAS Number: 1131587-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR