2-(Pyridin-3-yl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A503891-1G A503891-250MG A503891-5G
77034-514EA 21 USD
77034-514 77034-516 77034-518
2-(Pyridin-3-yl)ethanamine hydrochloride 95%
2-(Pyridin-3-yl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
MDL Number: MFCD09261148
CAS Number: 84359-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR