tert-Butyl 2,4-dioxo-3,8-diazaspiro[4.5]decane-8-carboxylate 98%

Supplier: Ambeed
Warning

A503740-100MG A503740-1G A503740-250MG
77034-508EA 49 USD
77034-508 77034-510 77034-512
tert-Butyl 2,4-dioxo-3,8-diazaspiro[4.5]decane-8-carboxylate 98%
tert-Butyl 2,4-dioxo-3,8-diazaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₄
MDL Number: MFCD12195867
CAS Number: 752234-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR