(S)-2-(((Benzyloxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Z-Asn(Trt)-OH, N-Cbz-N'-trityl-L-asparagine

A502681-100G A502681-10G A502681-1G A502681-25G A502681-5G
77004-700EA 103 USD
77004-700 77004-702 77004-704 77004-706 77004-708
(S)-2-(((Benzyloxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoic acid 98%
(S)-2-(((Benzyloxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoic acid

Precaución: for Research Use Only

Formula: C₃₁H₂₈N₂O₅
MDL Number: MFCD00153326
CAS Number: 132388-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR