2,3-Dichloroquinoxaline-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A502468-1G A502468-250MG
76991-268EA 391 USD
76991-268 77891-022
2,3-Dichloroquinoxaline-5-carboxylic acid 95%
2,3-Dichloroquinoxaline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₄Cl₂N₂O₂
MDL Number: MFCD22571787
CAS Number: 933726-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR