2,6-Dibromo-4-cyanophenyl octanoate 95%
Supplier: Ambeed
|
Danger
|
A502116-1G
A502116-5G
77004-696EA
46
USD
77004-696
77004-698
2,6-Dibromo-4-cyanophenyl octanoate 95%
2,6-Dibromo-4-cyanophenyl octanoate
Precaución: for Research Use Only
Formula:
C₁₅H₁₇Br₂NO₂ |
MDL Number:
MFCD00055484 CAS Number: 1689-99-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...