Boc-(R)-2-Cyclopentylglycine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

A501659-100MG A501659-1G A501659-250MG
77004-690EA 21 USD
77004-690 77004-692 77004-694
Boc-(R)-2-Cyclopentylglycine 97%
Boc-(R)-2-Cyclopentylglycine

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.299 g/mol
MDL Number: MFCD00671402
CAS Number: 156881-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR