4-(Methylsulfonyl)pyridine 97%

Supplier: Ambeed
Warning

A501506-100MG A501506-250MG A501506-1G
76991-258EA 78.51 USD
76991-258 76991-260 77613-582
4-(Methylsulfonyl)pyridine 97%
4-(Methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
MDL Number: MFCD18802769
CAS Number: 17075-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR