Isoquinolin-5-amine hydrochloride 98%

Supplier: Ambeed
Warning

A501493-100MG A501493-1G A501493-250MG
76991-252EA 12 USD
76991-252 76991-254 76991-256
Isoquinolin-5-amine hydrochloride 98%
Isoquinolin-5-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₉ClN₂
MDL Number: MFCD00035269
CAS Number: 152814-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR