1-Bromo-4-isopropoxybenzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-isopropoxylbenzene

A501486-1G A501486-250MG A501486-5G
77034-502EA 21.22 USD
77034-502 77034-504 77034-506
1-Bromo-4-isopropoxybenzene 95%
1-Bromo-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.1 g/mol
MDL Number: MFCD00089972
CAS Number: 6967-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR