4-((2-(Bromomethyl)phenyl)sulfonyl)morpholine 95%

Supplier: Ambeed
Danger

Synonyms: 2-[(Morpholin-4-yl)sulphonyl]benzyl bromide

A501301-1G A501301-250MG
76991-248EA 202 USD
76991-248 76991-250
4-((2-(Bromomethyl)phenyl)sulfonyl)morpholine 95%
4-((2-(Bromomethyl)phenyl)sulfonyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₃S
MDL Number: MFCD09879984
CAS Number: 941717-06-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR