(4-((Phenylamino)methyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A500947-1G
A500947-250MG
A500947-5G
76991-242EA
65
USD
76991-242
76991-244
76991-246
(4-((Phenylamino)methyl)phenyl)boronic acid 98%
(4-((Phenylamino)methyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BNO₂ |
MDL Number:
MFCD10699630 CAS Number: 690957-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...