2-Oxo-2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

A500804-100MG A500804-1G A500804-5G A500804-25G
77017-222EA 14 USD
77017-222 77017-224 77017-226 77891-018
2-Oxo-2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester 98%
2-Oxo-2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
MDL Number: MFCD01318910
CAS Number: 141293-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR