(2R,3S,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 98%

Supplier: Ambeed
Warning

Synonyms: DMT-dC(bz) Phosphoramidite

A500639-250MG A500639-5G A500639-25G A500639-1G A500639-100G
77034-498EA 7 USD
77034-498 77034-500 77572-586 77602-158 77882-239
(2R,3S,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 98%
(2R,3S,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₄₆H₅₂N₅O₈P
MW: 833.90 g/mol
MDL Number: MFCD00036315
CAS Number: 102212-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR