(6-Aminopyridin-2-yl)(1-methylpiperidin-4-yl)methanone 98%

Supplier: Ambeed
Warning

A500355-250MG A500355-1G A500355-5G
76991-240EA 59.42 USD
76991-240 77572-584 77684-319
(6-Aminopyridin-2-yl)(1-methylpiperidin-4-yl)methanone 98%
(6-Aminopyridin-2-yl)(1-methylpiperidin-4-yl)methanone

Precaución: For research use only.

Formula: C₁₂H₁₇N₃O
MW: 219.28 g/mol
MDL Number: MFCD22575148
CAS Number: 613678-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR