Pramipexole dihydrochloride monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride, (S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride hydrate

A500308-1G A500308-250MG A500308-25G A500308-5G
76991-232EA 31 USD
76991-232 76991-234 76991-236 76991-238
Pramipexole dihydrochloride monohydrate 99%
Pramipexole dihydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₂₁Cl₂N₃OS
MDL Number: MFCD02183927
CAS Number: 191217-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR