Ethyl-4-formyl-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A499847-100MG A499847-1G A499847-250MG A499847-5G
77034-490EA 11 USD
77034-490 77034-492 77034-494 77673-304
Ethyl-4-formyl-1H-pyrazole-5-carboxylate 98%
Ethyl-4-formyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD03768340
CAS Number: 179692-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR