4-Bromo-3-chloro-5-(trifluoromethyl)aniline 98+%

Supplier: Ambeed
Warning

A499777-1G A499777-250MG A499777-5G
77017-216EA 26 USD
77017-216 77017-218 77017-220
4-Bromo-3-chloro-5-(trifluoromethyl)aniline 98+%
4-Bromo-3-chloro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃N
MW: 274.47 g/mol
MDL Number: MFCD09033154
CAS Number: 914225-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR