Methyl-4-cyano-3-hydroxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-cyano-3-hydroxybenzoate

A499600-100MG A499600-250MG
77017-212EA 42 USD
77017-212 77017-214
Methyl-4-cyano-3-hydroxybenzoate 95%
Methyl-4-cyano-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD09836734
CAS Number: 6520-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR