(6-Chloro-1H-benzimidazol-2-yl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: (6-Chloro-1H-benzo[d]imidazol-2-yl)methanol

A499282-10G A499282-1G A499282-25G
77034-484EA 120 USD
77034-484 77034-486 77034-488
(6-Chloro-1H-benzimidazol-2-yl)methanol 95%
(6-Chloro-1H-benzimidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O
MW: 182.61 g/mol
MDL Number: MFCD09971879
CAS Number: 6953-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR