2,5-Dioctyl-3,6-di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%

Supplier: Ambeed
Warning

A498915-100MG A498915-1G A498915-250MG A498915-5G
76991-220EA 12 USD
76991-220 76991-222 76991-224 76991-226
2,5-Dioctyl-3,6-di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%
2,5-Dioctyl-3,6-di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂O₂S₂
MDL Number: MFCD26743580
CAS Number: 1057401-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR