2-Bromo-7-iodo-9,9-dimethyl-9H-fluorene 95%

Supplier: Ambeed
Warning

A498670-100G A498670-10G A498670-5G
76991-214EA 96 USD
76991-214 76991-216 76991-218
2-Bromo-7-iodo-9,9-dimethyl-9H-fluorene 95%
2-Bromo-7-iodo-9,9-dimethyl-9H-fluorene

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrI
MDL Number: MFCD12024277
CAS Number: 319906-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR