4-Methylquinolin-3-ol 97%

Supplier: Ambeed
Warning

A498635-1G A498635-5G
76991-210EA 251.44 USD
76991-210 76991-212
4-Methylquinolin-3-ol 97%
4-Methylquinolin-3-ol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD18414789
CAS Number: 6220-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR