1-(Bromomethyl)-2-fluoro-4-iodobenzene 97%

Supplier: Ambeed
Danger

A498082-100MG A498082-1G A498082-250MG
76991-204EA 42.44 USD
76991-204 76991-206 76991-208
1-(Bromomethyl)-2-fluoro-4-iodobenzene 97%
1-(Bromomethyl)-2-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MW: 314.92 g/mol
MDL Number: MFCD11847124
CAS Number: 85510-81-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR