1-Boc-6-aminoindole 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-amino-1H-indole-1-carboxylate

A497978-1G A497978-250MG A497978-5G
77004-670EA 89.12 USD
77004-670 77004-672 77004-674
1-Boc-6-aminoindole 98%
1-Boc-6-aminoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD04114745
CAS Number: 219508-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR