6-(2-Furyl)pyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A497967-1G A497967-250MG
77034-478EA 33 USD
77034-478 77034-480
6-(2-Furyl)pyridazin-3(2H)-one 95%
6-(2-Furyl)pyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD08289637
CAS Number: 38530-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR