γ-(N-Morpholino)propanesulphonic acid (MOPS) 97%

Supplier: Ambeed
Warning

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

A497711-1000G A497711-100G A497711-25G A497711-500G
77034-464EA 85 USD
77034-464 77034-466 77034-468 77034-470
γ-(N-Morpholino)propanesulphonic acid (MOPS) 97%
γ-(N-Morpholino)propanesulphonic acid (MOPS)

Precaución: for Research Use Only

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Flash Pt: 110
MDL Number: MFCD00006183
CAS Number: 1132-61-2
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR