tert-Butyl 5-amino-3,3-difluoropiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A496473-100MG A496473-1G A496473-250MG
77017-194EA 116.7 USD
77017-194 77017-196 77017-198
tert-Butyl 5-amino-3,3-difluoropiperidine-1-carboxylate 98%
tert-Butyl 5-amino-3,3-difluoropiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈F₂N₂O₂
MW: 236.26 g/mol
MDL Number: MFCD18433745
CAS Number: 1258638-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR