Cyclopropanesulfinic acid sodium salt 90%

Supplier: Ambeed
Warning

A496369-1G A496369-250MG A496369-5G A496369-25G
77004-658EA 28 USD
77004-658 77004-660 77004-662 77882-232
Cyclopropanesulfinic acid sodium salt 90%
Cyclopropanesulfinic acid sodium salt

Precaución: for Research Use Only

Formula: C₃H₅NaO₂S
MDL Number: MFCD08276208
CAS Number: 910209-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR