N-Acetyl 5-fluoro-2-methylaniline 98%

Supplier: Ambeed
Warning

A495861-1G A495861-25G A495861-5G
77004-652EA 5 USD
77004-652 77004-654 77004-656
N-Acetyl 5-fluoro-2-methylaniline 98%
N-Acetyl 5-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₀FNO
MDL Number: MFCD00045064
CAS Number: 366-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR