2-Chloro-5-fluoro-6-methoxynicotinic acid 98%

Supplier: Ambeed
Danger

A495594-100MG A495594-1G A495594-250MG A495594-5G
77004-644EA 19 USD
77004-644 77004-646 77004-648 77004-650
2-Chloro-5-fluoro-6-methoxynicotinic acid 98%
2-Chloro-5-fluoro-6-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₃
MDL Number: MFCD12025843
CAS Number: 943025-86-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR