1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(6-Chloro-5-nitro-4-pyrimidinyl)piperazine

A495106-1G A495106-250MG A495106-5G
77034-446EA 176.11 USD
77034-446 77034-448 77034-450
1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine 95%
1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClN₅O₄
MW: 343.7700 g/mol
MDL Number: MFCD11040204
CAS Number: 147539-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR