4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperazine-1-carboxylate
A495105-1G
A495105-250MG
A495105-5G
77017-182EA
66
USD
77017-182
77017-184
77017-186
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 96%
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₂₂H₃₃BN₂O₅ |
MDL Number:
MFCD08706021 CAS Number: 864754-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...