Adamantane-1,3-dicarbonitrile 95%

Supplier: Ambeed
Warning

A495044-100MG A495044-1G A495044-250MG A495044-5G
76991-184EA 10 USD
76991-184 76991-186 76991-188 76991-190
Adamantane-1,3-dicarbonitrile 95%
Adamantane-1,3-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MDL Number: MFCD00233041
CAS Number: 62472-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR