(4-Chlorobenzyl)triphenylphosphonium chloride 97%

Supplier: Ambeed
Warning

A494980-100G A494980-1G A494980-25G A494980-5G
77034-438EA 245 USD
77034-438 77034-440 77034-442 77034-444
(4-Chlorobenzyl)triphenylphosphonium chloride 97%
(4-Chlorobenzyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₅H₂₁Cl₂P
MDL Number: MFCD00041533
CAS Number: 1530-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR