1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: TIPS-EBX, 1-((Tris(1-methylethyl)silyl)ethynyl)-1,2-benziodoxol-3(1H)-one

A494645-100MG A494645-1G A494645-250MG
76991-178EA 56.23 USD
76991-178 76991-180 76991-182
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98%
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one

Precaución: for Research Use Only

Formula: C₁₈H₂₅IO₂Si
MW: 428.39 g/mol
MDL Number: MFCD18632570
CAS Number: 181934-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR