4-([2,2':6',2''-Terpyridin]-4'-yl)aniline 97%

Supplier: Ambeed
Warning

A494540-100MG A494540-250MG A494540-5G
77017-176EA 38.2 USD
77017-176 77017-178 77017-180
4-([2,2':6',2''-Terpyridin]-4'-yl)aniline 97%
4-([2,2':6',2''-Terpyridin]-4'-yl)aniline

Precaución: for Research Use Only

Formula: C₂₁H₁₆N₄
MW: 324.37 g/mol
MDL Number: MFCD06796989
CAS Number: 178265-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR